Cyclododecanol ≥99%
Supplier: Thermo Fisher Scientific
|
|
B23486.14
B23486.22
B23486.36
B23486.14EA
34.7
EUR
B23486.14
B23486.22
B23486.36
Cyclododecanol ≥99%
Cyclododecanol
Purity: 99%
CAS No.: 1724-39-6
Molecular Formula: C12H24O
Formula Weight: 184.32
Flash Point: 138°(280°F)
Melting Point: 77-79°
UN No.: 3077
MDL No.: MFCD00003717
Formula:
C₁₂H₂₄O Boiling Pt: 272.7 °C (1013 hPa) Melting Pt: 77…79 °C Flash Pt: 138 °C (280 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003717 CAS Number: 1724-39-6 EINECS: 217-031-2 UN: 3077 ADR: 9,III |