Chloro(diethyl)phosphine 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Diethylchlorophosphine
350310010 350310050
ACRO350310010EA 82.5 EUR
ACRO350310010 ACRO350310050
Chloro(diethyl)phosphine 95%
Chloro(diethyl)phosphine
Formula: C₄H₁₀ClP
MW: 124.55 g/mol
Boiling Pt: 131…132 °C (760 mmHg)
Density: 1.023 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00013621
CAS Number: 686-69-1
EINECS: 211-689-4
UN: 3129
ADR: 4.3 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
NMR ≥94 %