4-Methylphthalic anhydride 96%
Supplier: Thermo Fisher Scientific
Warning
414790250 414790050 414791000
ACRO414790250EA 45.1 EUR
ACRO414790250 ACRO414790050 ACRO414791000
4-Methylphthalic anhydride 96%
4-Methylphthalic anhydride
Formula: C₉H₆O₃
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00041856
CAS Number: 19438-61-0
EINECS: 243-073-6
Order Now
Specification Test Results
Appearance White to yellow to light orange Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 88 to 93 °C
GC ≥95 %
Titration by coupling ≥95 %