Calcium trifluoromethanesulphonate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Calcium trifluoromethanesulphonate, Calcium triflate

40134.09 40134.18 40134.22
40134.09EA 126 EUR
40134.09 40134.18 40134.22
Calcium trifluoromethanesulphonate ≥99%
Calcium trifluoromethanesulphonate

Purity: 99% min CAS No.: 55120-75-7 Molecular Formula: C2CaF6O6S2 Formula Weight: 338.21 Physical Form: Powder Appearance: White Melting Point: dec. MDL No.: MFCD00143911

Formula: C₂CaF₆O₆S₂
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00143911
CAS Number: 55120-75-7
EINECS: 232-188-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR