p-Tolyltrichlorosilane ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
B21728.06
B21728.14
B21728.06EA
37.8
EUR
B21728.06
B21728.14
p-Tolyltrichlorosilane ≥97%
p-Tolyltrichlorosilane
Purity: 97%
CAS No.: 701-35-9
Molecular Formula: C7H7Cl3Si
Formula Weight: 225.58
Flash Point: 98°(208°F)
Refractive Index: 1.5240
Odour: Irritating
Boiling Point: 218-220°
Beilstein Reference: 1866557
UN No.: 2987
MDL No.: MFCD00045131
Formula:
C₇H₇Cl₃Si Boiling Pt: 218…220 °C Density: 1.280 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045131 CAS Number: 701-35-9 EINECS: 211-854-0 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...