2,6-Dinitrobenzonitrile ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L12624.03
L12624.06
L12624.03EA
96
EUR
L12624.03
L12624.06
2,6-Dinitrobenzonitrile ≥98%
2,6-Dinitrobenzonitrile
Purity: 98+%
CAS No.: 35213-00-4
Molecular Formula: C7H3N3O4
Formula Weight: 193.12
Melting Point: 146-150°
Beilstein Reference: 2215516
UN No.: 3439
MDL No.: MFCD00041246
Formula:
C₇H₃N₃O₄ Melting Pt: 146…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041246 CAS Number: 35213-00-4 UN: 3439 ADR: 6.1,III |