Terpineol, mixture of isomers
Supplier: Roth Carl
8672.1 8672.2
ROTH8672.1EA 0 EUR
ROTH8672.1 ROTH8672.2
Terpineol, mixture of isomers
Terpineol, mixture of isomers
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00166983
CAS Number: 8000-41-7
EINECS: 232-268-1