Ethyl-3-bromopropionate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethyl 3-bromopropanoate

M-Clarity™ quality level = MQ200

128163-100G 128163-25G
SIAL128163-100GEA 94.3 EUR
SIAL128163-100G SIAL128163-25G
Ethyl-3-bromopropionate, Sigma-Aldrich®
Ethyl 3-bromopropionate
Formula: C₅H₉BrO₂
MDL Number: MFCD00000251
CAS Number: 539-74-2
EINECS: 208-724-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR