Pyridinium tribromide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide
M-Clarity™ quality level = MQ200
133248-100G
133248-25G
133248-500G
SIAL133248-100GEA
81.2
EUR
SIAL133248-100G
SIAL133248-25G
SIAL133248-500G
Pyridinium tribromide, Sigma-Aldrich®
Pyridinium tribromide
Formula:
C₅H₆Br₃N MW: 319.82 g/mol |
MDL Number:
MFCD00013223 CAS Number: 39416-48-3 EINECS: 254-446-8 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...