Dicyclohexyl phthala, Sigma-Aldrich®

Supplier: Merck

Synonyms: Dicyclohexyl phthalate

M-Clarity™ quality level = MQ100

306150-250G 306150-5G
SIAL306150-250GEA 77.7 EUR
SIAL306150-250G SIAL306150-5G
Dicyclohexyl phthala, Sigma-Aldrich®
Dicyclohexyl phthala
Formula: C₂₀H₂₆O₄
MDL Number: MFCD00003849
CAS Number: 84-61-7
EINECS: 201-545-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR