Tin(IV) acetate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

345172-10G 345172-2G
SIAL345172-10GEA 587 EUR
SIAL345172-10G SIAL345172-2G
Tin(IV) acetate, Sigma-Aldrich®
Tin (IV) acetate
Formula: C₈H₁₂O₈Sn
MDL Number: MFCD00014976
CAS Number: 2800-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR