2-Bromo-5-formylpyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde
M-Clarity™ quality level = MQ100
596280-1G 596280-25G 596280-5G
SIAL596280-1GEA 29.3 EUR
SIAL596280-1G SIAL596280-25G SIAL596280-5G
2-Bromo-5-formylpyridine, Sigma-Aldrich®
2-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
MDL Number: MFCD04115419
CAS Number: 149806-06-4