1-(4-Methoxybenzyl)piperazine, Sigma-Aldrich®

Supplier: Merck

646156-1G 646156-5G
SIAL646156-1GEA 87.7 EUR
SIAL646156-1G SIAL646156-5G
1-(4-Methoxybenzyl)piperazine, Sigma-Aldrich®
1-(4-Methoxybenzyl)piperazine
Formula: C₁₂H₁₈N₂O
MDL Number: MFCD01075236
CAS Number: 21867-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR