4-Methylcyclohexanone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13186.14
A13186.22
A13186.14EA
35.7
EUR
A13186.14
A13186.22
4-Methylcyclohexanone ≥98%
4-Methylcyclohexanone
Purity: 98+%
CAS No.: 589-92-4
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 48°(118°F)
Refractive Index: 1.4450
Melting Point: -41°
Boiling Point: 169-171°
Beilstein Reference: 506746
UN No.: 2297
MDL No.: MFCD00001643
Formula:
C₇H₁₂O MW: 112.17 g/mol Boiling Pt: 169…171 °C Melting Pt: –41 °C Density: 0.914 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001643 CAS Number: 589-92-4 EINECS: 209-665-3 UN: 2297 ADR: 3 III |