Bicyclohexyl ≥98%
Supplier: Thermo Fisher Scientific
Warning
|
A13481.09
A13481.18
A13481.09EA
40.4
EUR
A13481.09
A13481.18
Bicyclohexyl ≥98%
Bicyclohexyl
Purity: 98%
CAS No.: 92-51-3
Molecular Formula: C12H22
Formula Weight: 166.31
Flash Point: 92°(198°F)
Refractive Index: 1.478
Melting Point: 3-4°
Boiling Point: 226-228°
Beilstein Reference: 1848266
MDL No.: MFCD00003815
Formula:
C₁₂H₂₂ Boiling Pt: 226…228 °C Melting Pt: 3…4 °C Density: 0.864 Flash Pt: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003815 CAS Number: 92-51-3 EINECS: 202-161-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...