1-Methylcyclohexene 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
L15014.06
L15014.14
L15014.06EA
31.7
EUR
L15014.06
L15014.14
1-Methylcyclohexene 96%
1-Methylcyclohexene
Purity: 96%
CAS No.: 591-49-1
Molecular Formula: C7H12
Formula Weight: 96.17
Flash Point: -1°(30°F)
Refractive Index: 1.4500
Boiling Point: 108-112°
Beilstein Reference: 1304483
UN No.: 3295
MDL No.: MFCD00001548
Formula:
C₇H₁₂ Boiling Pt: 108…112 °C Melting Pt: –120.4 °C Density: 0.810 Flash Pt: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001548 CAS Number: 591-49-1 EINECS: 209-718-0 UN: 3295 ADR: 3,II |