Lithium orthoarsenate ≥99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Lithium arsenate
39350.06 39350.14
39350.06EA 56.3 EUR
39350.06 39350.14
Lithium orthoarsenate ≥99%
Lithium orthoarsenate
Purity: 99% CAS No.: 13478-14-3 Molecular Formula: Li3AsO4 Formula Weight: 159.74 Physical Form: Powder Density: 3.07 Solubility: Soluble in dilute acetic acid. Insoluble in water UN No.: 1557 MDL No.: MFCD00053998
Formula: AsLi₃O₄
MW: 159.74 g/mol
Density: 3.07
Storage Temperature: Ambient
MDL Number: MFCD00053998
CAS Number: 13478-14-3
EINECS: 236-773-8
UN: 1557
ADR: 6.1 II