di-Sodium tetraborate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Borax, Sodium tetraborate
M-Clarity™ quality level = MQ100
229946-100G 229946-25G 229946-5G
SIAL229946-100GEA 232 EUR
SIAL229946-100G SIAL229946-25G SIAL229946-5G
di-Sodium tetraborate, Sigma-Aldrich®
di-Sodium tetraborate
Formula: Na₂B₄O₇
MW: 201.22 g/mol
Boiling Pt: 1575
Melting Pt: 742
Density: 2.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 12,08733