Cesium chloride 99.999% (trace metals basis), extra pure
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Cesium Chloride
192810100 192810500 192812500
ACRO192810100EA 56 EUR
ACRO192810100 ACRO192810500 ACRO192812500
Cesium chloride 99.999% (trace metals basis), extra pure
Cesium chloride
Formula: CsCl
Boiling Pt: 1290 °C (760 mmHg)
Melting Pt: 646 °C
Density: 3,7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021
Order Now
Specification Test Results
Appearance White Powder
Total trace metal impurities ≤0.001 %