VWR®, Filter Tips, Low Retention
Supplier: VWR Collection
732-1488EA 152 EUR
732-1488
VWR®, Filter Tips, Low Retention
Pipette Tips
Low retention tips made of transparent PP resins offering unsurpassed fluid retention performance and proven aerosol blocking with endotoxin-free, 10 μm porous, hydrophobic filter. Ideal for PCR and DNA manipulation.
  • Sustainable design based on virgin resins
  • With PE filter, sterile
  • Up to 10 times less fluid retention than standard tips
  • Sample can be completely recovered without cutting into tip
  • Lot-certified purity (free from protease, RNase/DNase, human DNA and endotoxin, non pyrogenic)
  • Tips and racks are autoclavable to 121 °C for a maximum of 15 minutes (dry cycle is not recommended)