VWR® All-In-One, Chairs, Alu
Supplier: VWR Collection
132-0746EA 442 EUR
132-0746 132-0745 132-0743 132-0744
VWR® All-In-One, Chairs, Alu
Chairs
The VWR® All-In-One Alu is the upgrade version of the All-In-One. All the chairs are designed with a polished aluminium 5 star base. The seat height adjustment allows the individual adjustment of the seat height depending on the user's seating habits, activity and height. Also, the seat inclination adjustment enables inclination of the entire seat up to 5°, providing supported active sitting – ensuring a vertical position of the pelvis, helping the spine to maintain its natural S shape.
  • SoftTouch PU foam, resistant against aggressive liquids, easy to clean and disinfect
  • Permanent contact backrest responds to any movement and supports dynamic sitting
  • Seat height adjustment by gas-lift
  • Ergonomically shaped backrest for ideal fitting of the back
Glides: The glides are wear-proof plastic and have a large surface area offering a high level of stability.
Castors: Dual-wheel castors with load-sensitive brakes have to be right for the floor. All chairs are supplied with soft castors for hard floors as standard. Hard castors for soft floors are also available as an option.
Stop&Go technology: For safety reasons chairs whose seat is more than 650 mm high must not be movable whilst in use. With innovative Stop&Go castors these chairs are only mobile when no one is sitting on them.
Certifications: Conform to security level S2, GMP.