Potassium fluoride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
60238-100G-F
60238-500G-F
SIAL60238-100G-FEA
72.8
EUR
SIAL60238-100G-F
SIAL60238-500G-F
Potassium fluoride, Sigma-Aldrich®
Potassium fluoride
Formula:
KF Boiling Pt: 1505 °C (1013 hPa) Melting Pt: 860 °C Density: 2.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 13,07714 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...