Lithium iodide, anhydrous ≥99.95% (metals basis)
Supplier: Thermo Fisher Scientific
|
Warning
|
Ampuled under argon
40666.14
40666.22
40666.36
40666.14EA
263
EUR
40666.14
40666.22
40666.36
Lithium iodide, anhydrous ≥99.95% (metals basis)
Lithium iodide
Purity: 99.95%
CAS No.: 10377-51-2
Molecular Formula: ILi
Formula Weight: 133.84
Physical Form: Crystalline/Powder, Ampouled under argon
Density: 4.076
Refractive Index: 1.955
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 449°
Boiling Point: 1170-1190°
Merck Reference: 14,5535
MDL No.: MFCD00011092
Formula:
ILi MW: 133.85 g/mol Boiling Pt: 1170…1190 °C Melting Pt: 449 °C Density: 4.076 Storage Temperature: Ambient |
MDL Number:
MFCD00011092 CAS Number: 10377-51-2 EINECS: 233-822-5 Merck Index: 14,5535 |