Potassium iodate ≥98%
Supplier: Thermo Fisher Scientific
Danger
A16162.0E A16162.22 A16162.36
A16162.0EEA 606 EUR
A16162.0E A16162.22 A16162.36
Potassium iodate ≥98%
Potassium iodate
Purity: 98% CAS No.: 7758-05-6 Molecular Formula: KIO3 Formula Weight: 214.00 Melting Point: 560° dec. Merck Reference: 14,7642 MDL No.: MFCD00011406
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560 °C (dec.)
Density: 3.89
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1 II
Merck Index: 13,07726