Sodium iodate ≥99%
Supplier: Thermo Fisher Scientific
Danger
40135.22 40135.36
40135.22EA 63.4 EUR
40135.22 40135.36
Sodium iodate ≥99%
Sodium iodate
Purity: 99% min CAS No.: 7681-55-2 Molecular Formula: NaIO3 Formula Weight: 197.89 Physical Form: Crystalline powder Density: 4.28 Appearance: White Melting Point: dec. Merck Reference: 14,8630 UN No.: 1479 MDL No.: MFCD00003533
Formula: INaO₃
MW: 197.89 g/mol
Melting Pt: Dec.
Density: 4.28
Storage Temperature: Ambient
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1 II
Merck Index: 14,8630
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM