Cesium nitrate ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Nitric acid cesium salt
38617.14
38617.22
38617.14EA
238
EUR
38617.14
38617.22
Cesium nitrate ≥99.999% (metals basis)
Cesium nitrate
Purity: 99.999%
CAS No.: 7789-18-6
Molecular Formula: CsNO3
Formula Weight: 194.91
Physical Form: Crystalline
Density: 3.68
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 414°
Boiling Point: dec.
Merck Reference: 14,2015
UN No.: 1451
MDL No.: MFCD00010963
Formula:
CsNO₃ Boiling Pt: Dec. Melting Pt: 414 °C Density: 3.68 Storage Temperature: Ambient |
MDL Number:
MFCD00010963 CAS Number: 7789-18-6 EINECS: 232-146-8 UN: 1451 ADR: 5.1,III Merck Index: 14,2015 |