Water, UltraPure for molecular biology, sterile filtered
Supplier: Rockland Immunochemicals
MB-010-1000 MB-010-0100
ROCKMB-010-1000EA 192 EUR
ROCKMB-010-1000 ROCKMB-010-0100
Water, UltraPure for molecular biology, sterile filtered
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098