Water
Supplier: Roth Carl
Synonyms: Hydrogen oxide
3478.1 3478.2 3478.3
ROTH3478.1EA 0 EUR
ROTH3478.1 ROTH3478.2 ROTH3478.3
Water
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098