Sodium hydroxide, Sigma-Aldrich®
Supplier: Merck
Danger
71687-100G 71687-500G
SIAL71687-100GEA 34.1 EUR
SIAL71687-100G SIAL71687-500G
Sodium hydroxide, Sigma-Aldrich®
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627