Disodium hydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
71636-1KG 71636-250G
SIAL71636-1KGEA 243 EUR
SIAL71636-1KG SIAL71636-250G
Disodium hydrogen phosphate, Sigma-Aldrich®
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805