Sodium thiocyanate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
467871-50G 467871-250G
SIAL467871-50GEA 101 EUR
SIAL467871-50G SIAL467871-250G
Sodium thiocyanate, Sigma-Aldrich®
Sodium thiocyanate
Formula: NaSCN
Melting Pt: 287 °C
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS Number: 540-72-7
EINECS: 208-754-4
Merck Index: 13,09401