Sodium sulphate, anhydrous ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
87611.06
87611.14
87611.22
87611.06EA
83.4
EUR
87611.06
87611.14
87611.22
Sodium sulphate, anhydrous ≥99.99% (metals basis)
Sodium sulphate
Purity: 99.99%
CAS No.: 7757-82-6
Molecular Formula: Na2O4S
Formula Weight: 142.04
Physical Form: Powder/chunks
Density: 2.680
Refractive Index: 1.4840
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 884°
Merck Reference: 14,8680
MDL No.: MFCD00003504
Formula:
Na₂SO₄ Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 884 °C Density: 2.680 Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 14,8680 |