Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR Chemicals
Warning
1B1110-1KGEA 284 EUR
1B1110-1KG 1B1110-2.5KG 1B1110-500G
Calcium chloride, anhydrous ≥96% ACS
Calcium chloride
Formula: CaCl₂
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659
Order Now
Specification Test Results
Purity >96%
Titrable Free Base ≤0.006 meq/g