Furfurylamine 99+%
Supplier: Thermo Fisher Scientific
Danger
119800010 119800050 119802500
ACRO119800010EA 219 EUR
ACRO119800010 ACRO119800050 ACRO119802500
Furfurylamine 99+%
Furfurylamine
Formula: C₅H₇NO
Boiling Pt: 145…146 °C (761 mmHg)
Melting Pt: –70 °C
Density: 1.09 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00003258
CAS Number: 617-89-0
EINECS: 210-536-9
UN: 2526
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
GC ≥99.0 %
Titration with HClO4 ≥99.0 %