Sodium benzenesulphinate ≥98%

Supplier: Thermo Fisher Scientific

A12118.14 A12118.22 A12118.36
A12118.14EA 27.4 EUR
A12118.14 A12118.22 A12118.36
Sodium benzenesulphinate ≥98%
Sodium benzenesulphinate

Purity: 98% CAS No.: 873-55-2 Molecular Formula: C6H5NaO2S Formula Weight: 164.16 Melting Point: 300° Beilstein Reference: 3598405

Formula: C₆H₅NaO₂S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013135
CAS Number: 873-55-2
EINECS: 212-842-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR