Phenoxyacetamide

Supplier: Thermo Scientific

SPB04334.1 SPB04334.10 SPB04334.50
MAYBSPB04334.1EA 0 EUR
MAYBSPB04334.1 MAYBSPB04334.10 MAYBSPB04334.50
Phenoxyacetamide
Phenoxyacetamide
Formula: C₈H₉NO₂
MDL Number: MFCD00017140
CAS Number: 621-88-5
EINECS: 210-713-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR