1,3-Benzenedisulphonyl chloride

Supplier: Thermo Scientific

TL00307.1 TL00307.10 TL00307.50
MAYBTL00307.1EA 0 EUR
MAYBTL00307.1 MAYBTL00307.10 MAYBTL00307.50
1,3-Benzenedisulphonyl chloride
1,3-Benzenedisulphonyl chloride
Formula: C₆H₄Cl₂O₄S₂
Boiling Pt: 145 °C (1 mmHg)
Melting Pt: 63 °C
MDL Number: MFCD00041879
CAS Number: 585-47-7
EINECS: 209-556-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR