Aluminium chloride, anhydrous ≥99%, powder
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Aluminum chloride
L18489.22 L18489.36
L18489.22EA 40.1 EUR
L18489.22 L18489.36
Aluminium chloride, anhydrous ≥99%, powder
Aluminium chloride
Purity: 99% CAS No.: 7446-70-0 Molecular Formula: AlCl3 Formula Weight: 133.34 Odour: Pungent pH: 2,4 Melting Point: 190° subl Merck Reference: 14,337 UN No.: 1726 MDL No.: MFCD00003422
Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180
Melting Pt: 190 °C (subl.)
Density: 2.44
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 14,337