1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers), Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,4-Cyclohexanedimethanol
M-Clarity™ quality level = MQ100
125598-100G 125598-17KG 125598-3KG 125598-500G
SIAL125598-100GEA 25.9 EUR
SIAL125598-100G SIAL125598-17KG SIAL125598-3KG SIAL125598-500G
1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers), Sigma-Aldrich®
1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers)
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00001512
CAS Number: 105-08-8
EINECS: 203-268-9