Myristoyl chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

185205-5ML 185205-100ML 185205-500ML
SIAL185205-5MLEA 41.3 EUR
SIAL185205-5ML SIAL185205-100ML SIAL185205-500ML
Myristoyl chloride, Sigma-Aldrich®
Myristoyl chloride
Formula: C₁₄H₂₇ClO
MDL Number: MFCD00000741
CAS Number: 112-64-1
EINECS: 203-994-6
UN: 3265
ADR: 8,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR