Ethylenediammonium dichloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethylenediamine dihydrochloride

M-Clarity™ quality level = MQ200

195804-100G 195804-500G
SIAL195804-100GEA 33.2 EUR
SIAL195804-100G SIAL195804-500G
Ethylenediammonium dichloride, Sigma-Aldrich®
Ethylenediammonium dichloride
Formula: C₂H₁₀Cl₂N₂
MW: 133.02 g/mol
MDL Number: MFCD00012524
CAS Number: 333-18-6
EINECS: 206-369-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR