Palladium 1% on activated carbon hydrogenation catalyst, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

205672-100G 205672-25G
SIAL205672-100GEA 621 EUR
SIAL205672-100G SIAL205672-25G
Palladium 1% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR