Platinum 1% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Supplier: Merck
Synonyms: Platinium
M-Clarity™ quality level = MQ100
205923-25G 205923-5G
SIAL205923-25GEA 208 EUR
SIAL205923-25G SIAL205923-5G
Platinum 1% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Platinum
Formula: Pt
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 1325
ADR: 4.1,II