Bis(cyclopentadienyl)zirconium hydridochloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Bis(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz's Reagent, Bis(cyclopentadienyl)zirconium chloride hydride

M-Clarity™ quality level = MQ100

223670-1G 223670-25G 223670-5G
SIAL223670-1GEA 41.8 EUR
SIAL223670-1G SIAL223670-25G SIAL223670-5G
Bis(cyclopentadienyl)zirconium hydridochloride, Sigma-Aldrich®
Bis(cyclopentadienyl)zirconium hydridochloride
Formula: C₁₀H₁₁ClZr
MW: 257.87 g/mol
MDL Number: MFCD02089401
CAS Number: 37342-97-5
EINECS: 253-479-5
UN: 3131
ADR: 4.3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR