N,N-Dimethylpropionamide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

252875-100G 252875-25G
SIAL252875-100GEA 291 EUR
SIAL252875-100G SIAL252875-25G
N,N-Dimethylpropionamide, Sigma-Aldrich®
N,N-Dimethylpropionamide
Formula: C₅H₁₁NO
MDL Number: MFCD00009301
CAS Number: 758-96-3
EINECS: 212-064-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR