Palladium 5% on activated carbon wetted with ca. 50% water hydrogenation catalyst, Sigma-Aldrich®
Supplier: Merck
Stabilization: Moistened with water
M-Clarity™ quality level = MQ100
276707-10G 276707-50G
SIAL276707-10GEA 171 EUR
SIAL276707-10G SIAL276707-50G
Palladium 5% on activated carbon wetted with ca. 50% water hydrogenation catalyst, Sigma-Aldrich®
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
ADR: 4.1,III