N-Methylallylamine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

317748-1G 317748-5G
SIAL317748-1GEA 30.5 EUR
SIAL317748-1G SIAL317748-5G
N-Methylallylamine, Sigma-Aldrich®
N-Methylallylamine
Formula: C₄H₉N
MDL Number: MFCD00008641
CAS Number: 627-37-2
EINECS: 210-996-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR