Palladium 10% on activated carbon wetted with ca. 50% water hydrogenation catalyst Degussa Type E101 NE/W, Sigma-Aldrich®
Supplier: Merck
Stabilization: Moistened with water
M-Clarity™ quality level = MQ200
330108-10G 330108-1G 330108-50G
SIAL330108-10GEA 377 EUR
SIAL330108-10G SIAL330108-1G SIAL330108-50G
Palladium 10% on activated carbon wetted with ca. 50% water hydrogenation catalyst Degussa Type E101 NE/W, Sigma-Aldrich®
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
ADR: 4.1,III