Phenylphosphonic dichloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Phenyldichlorophosphine oxide, Dichlorophenylphosphine oxide
M-Clarity™ quality level = MQ100
389560-100ML
389560-500ML
SIAL389560-100MLEA
87.9
EUR
SIAL389560-100ML
SIAL389560-500ML
Phenylphosphonic dichloride, Sigma-Aldrich®
Phenylphosphonic dichloride
Formula:
C₆H₅Cl₂OP |
MDL Number:
MFCD00002070 CAS Number: 824-72-6 EINECS: 212-534-3 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...