Dimethyl chlorophosphate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Chlorophosphoric acid dimethyl ester, Dimethyl phosphorochloridate
M-Clarity™ quality level = MQ100
437891-25ML
437891-5ML
SIAL437891-25MLEA
637
EUR
SIAL437891-25ML
SIAL437891-5ML
Dimethyl chlorophosphate, Sigma-Aldrich®
Dimethyl chlorophosphate
Formula:
C₂H₆ClO₃P |
MDL Number:
MFCD00117904 CAS Number: 813-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...